Assay Details
| Gene Symbol: LOC4326348 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765687.2 | XP_015621173.1 | 1 - 2 | 1128 | 66 | |
| GenBank mRNA | AK066090.1 | - | 2 - 3 | 255 | 66 | |
| AK121334.1 | - | 1 - 2 | 877 | 66 | ||
More Information
Set Membership: |
|

LOC4326348
Genomic Map