Assay Details
| Gene Symbol: LOC4326348 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765687.2 | XP_015621173.1 | 2 - 3 | 1206 | 70 | |
| GenBank mRNA | AK066090.1 | - | 3 - 4 | 333 | 70 | |
| AK121334.1 | - | 2 - 3 | 955 | 70 | ||
More Information
Set Membership: |
|

LOC4326348
Genomic Map