Assay Details
| Gene Symbol: LOC4326348 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765687.2 | XP_015621173.1 | 5 - 6 | 1734 | 88 | |
| GenBank mRNA | AK066090.1 | - | 6 - 7 | 861 | 88 | |
| AK121334.1 | - | 5 - 6 | 1483 | 88 | ||
More Information
Set Membership: |
|

LOC4326348
Genomic Map