Assay Details
| Gene Symbol: LOC4326348 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765687.2 | XP_015621173.1 | 6 - 7 | 1866 | 95 | |
| GenBank mRNA | AK066090.1 | - | 7 - 8 | 993 | 95 | |
| AK121334.1 | - | 6 - 7 | 1615 | 95 | ||
More Information
Set Membership: |
|

LOC4326348
Genomic Map