Assay Details
| Gene Symbol: LOC4326428 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773156.2 | XP_015628642.1 | 4 - 5 | 683 | 149 | |
| XM_015773165.2 | XP_015628651.1 | 4 - 5 | 683 | 149 | ||
| XM_015773175.2 | XP_015628661.1 | 4 - 5 | 683 | 149 | ||
| GenBank mRNA | AK241720.1 | - | 4 - 5 | 675 | 149 | |
More Information
Set Membership: |
|

LOC4326428
Genomic Map