Assay Details
| Gene Symbol: LOC4327848 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766581.2 | XP_015622067.1 | 2 - 3 | 923 | 79 | |
| GenBank mRNA | AK103516.1 | - | 2 - 3 | 918 | 79 | |
More Information
Set Membership: |
|

LOC4327848
Genomic Map