Assay Details
| Gene Symbol: LOC4327848 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766581.2 | XP_015622067.1 | 3 - 4 | 1052 | 78 | |
| GenBank mRNA | AK060870.1 | - | 592 | 78 | ||
| AK103516.1 | - | 3 - 4 | 1047 | 78 | ||
| AK104484.1 | - | 592 | 78 | |||
More Information
Set Membership: |
|

LOC4327848
Genomic Map