Assay Details
| Gene Symbol: LOC4327550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794238.2 | 1 - 2 | 108 | 61 | ||
| GenBank mRNA | AK072502.1 | - | 1 - 2 | 81 | 61 | |
| X78143.1 | - | 1 - 2 | 60 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the X78143.1 transcripts.
Set Membership: |
|

LOC4327550
Genomic Map