Assay Details
| Gene Symbol: LOC4327357 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788768.2 | XP_015644254.1 | 3 - 4 | 429 | 81 | |
| GenBank mRNA | AB026562.1 | - | 3 - 4 | 295 | 81 | |
| AK061784.1 | - | 3 - 4 | 332 | 81 | ||
| AK103541.1 | - | 3 - 4 | 399 | 81 | ||
More Information
Set Membership: |
|

LOC4327357
Genomic Map