Assay Details
| Gene Symbol: LOC4327357 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788768.2 | XP_015644254.1 | 9 - 10 | 862 | 131 | |
| GenBank mRNA | AB026562.1 | - | 9 - 10 | 728 | 131 | |
| AK061784.1 | - | 9 - 10 | 764 | 131 | ||
| AK103541.1 | - | 9 - 10 | 832 | 131 | ||
More Information
Set Membership: |
|

LOC4327357
Genomic Map