Assay Details
| Gene Symbol: LOC9271250 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784576.2 | XP_015640062.1 | 11 - 12 | 1482 | 67 | |
| GenBank mRNA | AK067520.1 | - | 11 - 12 | 1451 | 67 | |
More Information
Set Membership: |
|

LOC9271250
Genomic Map