Assay Details
| Gene Symbol: LOC4327974 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768401.2 | XP_015623887.1 | 1 | 1043 | 77 | |
| GenBank mRNA | AK062483.1 | - | 1 | 753 | 77 | |
More Information
Set Membership: |
|

LOC4327974
Genomic Map