Assay Details
| Gene Symbol: LOC4328073 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769909.2 | XP_015625395.1 | 1 - 2 | 92 | 73 | |
| GenBank mRNA | AK065094.1 | - | 1 - 2 | 89 | 73 | |
| AK099231.1 | - | 1 - 2 | 89 | 73 | ||
| AK099923.1 | - | 1 - 2 | 91 | 73 | ||
| JX393051.1 | - | 1 - 2 | 69 | 73 | ||
| JX972147.1 | - | 1 - 2 | 69 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015769909.2 transcripts.
Set Membership: |
|

LOC4328073
Genomic Map