Assay Details
| Gene Symbol: LOC4328073 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769909.2 | XP_015625395.1 | 2 - 3 | 890 | 103 | |
| GenBank mRNA | AK065094.1 | - | 2 - 3 | 887 | 103 | |
| AK068433.1 | - | 1 - 2 | 331 | 103 | ||
| AK099231.1 | - | 2 - 3 | 887 | 103 | ||
| AK099923.1 | - | 2 - 3 | 889 | 103 | ||
| JX393051.1 | - | 2 - 3 | 867 | 103 | ||
| JX972147.1 | - | 2 - 3 | 867 | 103 | ||
More Information
Set Membership: |
|

LOC4328073
Genomic Map