Assay Details
| Gene Symbol: LOC4328073 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769909.2 | XP_015625395.1 | 3 - 4 | 1494 | 69 | |
| GenBank mRNA | AK061923.1 | - | 1 - 2 | 96 | 69 | |
| AK065094.1 | - | 3 - 4 | 1491 | 69 | ||
| AK068433.1 | - | 3 - 4 | 720 | 69 | ||
| AK071061.1 | - | 1 - 2 | 632 | 69 | ||
| AK099231.1 | - | 3 - 4 | 1491 | 69 | ||
| AK099923.1 | - | 3 - 4 | 1493 | 69 | ||
| JX393051.1 | - | 3 - 4 | 1471 | 69 | ||
| JX972147.1 | - | 3 - 4 | 1471 | 69 | ||
More Information
Set Membership: |
|

LOC4328073
Genomic Map