Assay Details
| Gene Symbol: LOC4328109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769719.2 | XP_015625205.1 | 14 - 15 | 1705 | 65 | |
| GenBank mRNA | AK060220.1 | - | 6 - 7 | 545 | 65 | |
| AK103704.1 | - | 14 - 15 | 1706 | 65 | ||
More Information
Set Membership: |
|

LOC4328109
Genomic Map