Assay Details
| Gene Symbol: LOC112936036 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772229.2 | XP_015627715.1 | 2 - 3 | 1022 | 86 | |
| GenBank mRNA | AK073907.1 | - | 2 - 3 | 392 | 86 | |
| AK111055.1 | - | 2 - 3 | 372 | 86 | ||
More Information
Set Membership: |
|

LOC112936036
Genomic Map