Assay Details
| Gene Symbol: LOC112936036 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772229.2 | XP_015627715.1 | 3 - 4 | 1207 | 89 | |
| XM_015772230.2 | XP_015627716.1 | 3 - 4 | 1201 | 89 | ||
| GenBank mRNA | AK073907.1 | - | 3 - 4 | 577 | 89 | |
More Information
Set Membership: |
|

LOC112936036
Genomic Map