Assay Details
| Gene Symbol: LOC4328439 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767854.2 | XP_015623340.1 | 1 - 2 | 361 | 125 | |
| GenBank mRNA | AK070107.1 | - | 1 - 2 | 333 | 125 | |
| AK099345.1 | - | 1 - 2 | 309 | 125 | ||
| AK103039.1 | - | 1 - 2 | 272 | 125 | ||
| AK105887.1 | - | 1 - 2 | 334 | 125 | ||
More Information
Set Membership: |
|

LOC4328439
Genomic Map