Assay Details
| Gene Symbol: LOC4328439 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767854.2 | XP_015623340.1 | 2 - 3 | 425 | 74 | |
| GenBank mRNA | AK070107.1 | - | 2 - 3 | 397 | 74 | |
| AK099345.1 | - | 2 - 3 | 373 | 74 | ||
| AK103039.1 | - | 2 - 3 | 336 | 74 | ||
| AK105887.1 | - | 2 - 3 | 398 | 74 | ||
More Information
Set Membership: |
|

LOC4328439
Genomic Map