Assay Details
| Gene Symbol: LOC4328460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768527.2 | XP_015624013.1 | 3 - 4 | 840 | 69 | |
| XM_015768528.2 | XP_015624014.1 | 3 - 4 | 840 | 69 | ||
More Information
Set Membership: |
|

LOC4328460
Genomic Map