Assay Details
| Gene Symbol: LOC4328577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770102.2 | XP_015625588.1 | 1 - 2 | 207 | 66 | |
| GenBank mRNA | AK069748.1 | - | 1 - 2 | 74 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015770102.2 transcripts.
Set Membership: |
|

LOC4328577
Genomic Map