Assay Details
| Gene Symbol: LOC4328800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770473.2 | 4 - 5 | 544 | 120 | ||
| XM_015770474.2 | 4 - 5 | 544 | 120 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4328800| Gene Symbol: LOC4328800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770473.2 | 4 - 5 | 544 | 120 | ||
| XM_015770474.2 | 4 - 5 | 544 | 120 | |||
Set Membership: |
|