Assay Details
| Gene Symbol: LOC4328900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771402.1 | XP_015626888.1 | 1 - 2 | 110 | 145 | |
| GenBank mRNA | AK062577.1 | - | 1 - 2 | 116 | 145 | |
| AK099051.1 | - | 1 - 2 | 117 | 145 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015771402.1 transcripts.
Set Membership: |
|

LOC4328900
Genomic Map