Assay Details
| Gene Symbol: LOC4328900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771402.1 | XP_015626888.1 | 5 - 6 | 549 | 81 | |
| GenBank mRNA | AK062577.1 | - | 5 - 6 | 555 | 81 | |
| AK099051.1 | - | 5 - 6 | 556 | 81 | ||
More Information
Set Membership: |
|

LOC4328900
Genomic Map