Assay Details
Gene Symbol: LOC4329007 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015770843.2 | XP_015626329.1 | 7 - 8 | 1019 | 82 | |
GenBank mRNA | AK072052.1 | - | 7 - 8 | 999 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
