Assay Details
| Gene Symbol: LOC4329023 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771328.2 | XP_015626814.1 | 13 - 14 | 3528 | 115 | |
| XM_015771329.2 | XP_015626815.1 | 13 - 14 | 3475 | 115 | ||
| GenBank mRNA | AK068697.1 | - | 13 - 14 | 3477 | 115 | |
More Information
Set Membership: |
|

LOC4329023
Genomic Map