Assay Details
| Gene Symbol: LOC4329381 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772018.2 | XP_015627504.1 | 1 - 2 | 100 | 115 | |
| GenBank mRNA | AK102402.1 | - | 1 - 2 | 92 | 115 | |
| AK102757.1 | - | 1 - 2 | 92 | 115 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015772018.2 transcripts.
Set Membership: |
|

LOC4329381
Genomic Map