Assay Details
| Gene Symbol: LOC4329381 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772018.2 | XP_015627504.1 | 3 - 4 | 503 | 112 | |
| GenBank mRNA | AK102402.1 | - | 3 - 4 | 495 | 112 | |
| AK102757.1 | - | 3 - 4 | 495 | 112 | ||
| Gene Symbol: LOC4333453 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK241816.1 | - | 1054 | 112 | ||
More Information
Set Membership: |
|

LOC4329381
Genomic Map