Assay Details
| Gene Symbol: LOC4329476 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767433.2 | XP_015622919.1 | 2 - 3 | 488 | 73 | |
| XM_015767434.2 | XP_015622920.1 | 2 - 3 | 484 | 73 | ||
| GenBank mRNA | AK058947.1 | - | 2 - 3 | 468 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015767433.2 transcripts.
Set Membership: |
|

LOC4329476
Genomic Map