Assay Details
| Gene Symbol: LOC4329741 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770999.2 | XP_015626485.1 | 1 - 2 | 502 | 63 | |
| GenBank mRNA | AK121678.1 | - | 1 - 2 | 228 | 63 | |
More Information
Set Membership: |
|

LOC4329741
Genomic Map