Assay Details
Gene Symbol: LOC4350004 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK241764.1 | - | 1125 | 86 | ||
AK287873.1 | - | 1125 | 86 |
Gene Symbol: LOC4330472 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015770199.2 | XP_015625685.1 | 1 - 2 | 282 | 86 | |
GenBank mRNA | AK064357.1 | - | 1 - 2 | 254 | 86 | |
AK102318.1 | - | 1 - 2 | 202 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
