Assay Details
| Gene Symbol: LOC4350004 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK287873.1 | - | 883 | 110 | ||
| Gene Symbol: LOC4330472 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770199.2 | XP_015625685.1 | 3 - 4 | 524 | 110 | |
| GenBank mRNA | AK064357.1 | - | 3 - 4 | 496 | 110 | |
| AK102318.1 | - | 3 - 4 | 444 | 110 | ||
More Information
Set Membership: |
|

LOC4350004
Genomic Map