Assay Details
| Gene Symbol: LOC9272427 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768351.2 | XP_015623837.1 | 3 - 4 | 1620 | 110 | |
| GenBank mRNA | AK069555.1 | - | 4 - 5 | 1490 | 110 | |
More Information
Set Membership: |
|

LOC9272427
Genomic Map