Assay Details
| Gene Symbol: LOC4330683 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767284.2 | XP_015622770.1 | 4 - 5 | 805 | 120 | |
| GenBank mRNA | AK068712.1 | - | 4 - 5 | 793 | 120 | |
| AK071144.1 | - | 4 - 5 | 719 | 120 | ||
More Information
Set Membership: |
|

LOC4330683
Genomic Map