Assay Details
| Gene Symbol: LOC4330700 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771613.2 | XP_015627099.1 | 1 - 2 | 1426 | 69 | |
| GenBank mRNA | AK099634.1 | - | 1 - 2 | 1279 | 69 | |
| AK102343.1 | - | 1 - 2 | 1279 | 69 | ||
More Information
Set Membership: |
|

LOC4330700
Genomic Map