Assay Details
| Gene Symbol: LOC4330814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767555.2 | XP_015623041.1 | 3 - 4 | 696 | 80 | |
| GenBank mRNA | AK103885.1 | - | 3 - 4 | 656 | 80 | |
More Information
Set Membership: |
|

LOC4330814
Genomic Map