Assay Details
| Gene Symbol: LOC4330814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767555.2 | XP_015623041.1 | 4 - 5 | 860 | 94 | |
| GenBank mRNA | AK103885.1 | - | 4 - 5 | 820 | 94 | |
More Information
Set Membership: |
|

LOC4330814
Genomic Map