Assay Details
| Gene Symbol: LOC4330845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771117.2 | XP_015626603.1 | 2 - 3 | 827 | 69 | |
| GenBank mRNA | AB016065.1 | - | 2 - 3 | 798 | 69 | |
| AK069611.1 | - | 2 - 3 | 833 | 69 | ||
More Information
Set Membership: |
|

LOC4330845
Genomic Map