Assay Details
| Gene Symbol: LOC4330990 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767561.2 | XP_015623047.1 | 1 | 518 | 94 | |
| GenBank mRNA | AK062552.1 | - | 1 | 449 | 94 | |
More Information
Set Membership: |
|

LOC4330990
Genomic Map