Assay Details
| Gene Symbol: LOC4331066 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767668.2 | XP_015623154.1 | 1 - 2 | 336 | 90 | |
| GenBank mRNA | AK059822.1 | - | 1 - 2 | 324 | 90 | |
| AK099516.1 | - | 1 - 2 | 327 | 90 | ||
| AK104587.1 | - | 1 - 2 | 300 | 90 | ||
More Information
Set Membership: |
|

LOC4331066
Genomic Map