Assay Details
| Gene Symbol: LOC4331518 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777422.2 | XP_015632908.1 | 6 - 7 | 1202 | 85 | |
| GenBank mRNA | AK104997.2 | - | 6 - 7 | 1102 | 85 | |
| AK122069.1 | - | 6 - 7 | 1107 | 85 | ||
More Information
Set Membership: |
|

LOC4331518
Genomic Map