Assay Details
| Gene Symbol: LOC4331533 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773601.2 | XP_015629087.1 | 1 - 2 | 266 | 64 | |
| GenBank mRNA | AK059955.1 | - | 1 - 2 | 266 | 64 | |
| AY533123.1 | - | 1 - 2 | 158 | 64 | ||
More Information
Set Membership: |
|

LOC4331533
Genomic Map