Assay Details
| Gene Symbol: LOC4331790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772902.2 | XP_015628388.1 | 13 - 14 | 2393 | 100 | |
| XM_015772903.2 | XP_015628389.1 | 11 - 12 | 1805 | 100 | ||
| XM_015772904.2 | XP_015628390.1 | 11 - 12 | 1798 | 100 | ||
| GenBank mRNA | AK067994.1 | - | 13 - 14 | 2333 | 100 | |
More Information
Set Membership: |
|

LOC4331790
Genomic Map