Assay Details
| Gene Symbol: LOC4331790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772902.2 | XP_015628388.1 | 1 - 2 | 343 | 77 | |
| GenBank mRNA | AK067994.1 | - | 1 - 2 | 269 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015772902.2 transcripts.
Set Membership: |
|

LOC4331790
Genomic Map