Assay Details
| Gene Symbol: LOC4331790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772903.2 | XP_015628389.1 | 2 - 3 | 228 | 67 | |
| XM_015772904.2 | XP_015628390.1 | 2 - 3 | 221 | 67 | ||
| GenBank mRNA | AK067994.1 | - | 4 - 5 | 756 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015772903.2, XM_015772904.2 transcripts.
Set Membership: |
|

LOC4331790
Genomic Map