Assay Details
| Gene Symbol: LOC4331790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772902.2 | XP_015628388.1 | 5 - 6 | 1014 | 87 | |
| XM_015772903.2 | XP_015628389.1 | 3 - 4 | 426 | 87 | ||
| XM_015772904.2 | XP_015628390.1 | 3 - 4 | 419 | 87 | ||
| GenBank mRNA | AK067994.1 | - | 5 - 6 | 954 | 87 | |
More Information
Set Membership: |
|

LOC4331790
Genomic Map