Assay Details
| Gene Symbol: LOC4331790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772902.2 | XP_015628388.1 | 8 - 9 | 1574 | 70 | |
| XM_015772903.2 | XP_015628389.1 | 6 - 7 | 986 | 70 | ||
| XM_015772904.2 | XP_015628390.1 | 6 - 7 | 979 | 70 | ||
| GenBank mRNA | AK067994.1 | - | 8 - 9 | 1514 | 70 | |
More Information
Set Membership: |
|

LOC4331790
Genomic Map