Assay Details
| Gene Symbol: LOC4331814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774313.2 | XP_015629799.1 | 1 - 2 | 816 | 85 | |
| XM_015774314.2 | XP_015629800.1 | 1 - 2 | 820 | 85 | ||
| XM_015774315.2 | XP_015629801.1 | 1 - 2 | 821 | 85 | ||
More Information
Set Membership: |
|

LOC4331814
Genomic Map