Assay Details
| Gene Symbol: LOC4331814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774313.2 | XP_015629799.1 | 4 - 5 | 1046 | 96 | |
| XM_015774314.2 | XP_015629800.1 | 4 - 5 | 1050 | 96 | ||
| XM_015774315.2 | XP_015629801.1 | 4 - 5 | 1051 | 96 | ||
More Information
Set Membership: |
|

LOC4331814
Genomic Map